Learn More
Meso Scale Discovery UPLEX MSE IL17E/IL25 ASY
SDP
Catalog No. 5000079641
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-17E/IL-25 Assay Kit is a multiplex immunoassay that allows simultaneous measurement of mouse interleukin 17E (IL-17E) and interleukin 25 (IL-25) in serum plasma and cell culture supernatants The assay uses MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and an MSD read buffer The assay takes approximately 3 5 hours to complete after a 2-hour incubation
Catalog No. 50-000-79641 Supplier Meso Scale Discovery Supplier No. K152VZK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-17E/IL-25 Assay Kit is a multiplex immunoassay that allows simultaneous measurement of mouse interleukin 17E (IL-17E) and interleukin 25 (IL-25) in serum plasma and cell culture supernatants The assay uses MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and an MSD read buffer The assay takes approximately 3 5 hours to complete after a 2-hour incubation