Learn More
Meso Scale Discovery UPLEX MSE IL17E/IL25 ASY384
SDP
Catalog No. 5000081534
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-17E/IL-25 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 17E (IL-17E) and interleukin 25 (IL-25) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This 384-well plate assay allows for efficient and accurate quantification of IL-17E and IL-25 biomarkers The kit includes a plate coated with capture antibodies detection antibodies read buffer wash buffer calibrator blend and controls needed to run the assay
Catalog No. 50-000-81534 Supplier Meso Scale Discovery Supplier No. K252VZK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-17E/IL-25 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 17E (IL-17E) and interleukin 25 (IL-25) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This 384-well plate assay allows for efficient and accurate quantification of IL-17E and IL-25 biomarkers The kit includes a plate coated with capture antibodies detection antibodies read buffer wash buffer calibrator blend and controls needed to run the assay