Learn More
Meso Scale Discovery UPLEX MSE IL1B ASY384well
SDP
Catalog No. 5000079339
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-1beta 384-well Assay Kit is a multiplex immunoassay for measuring mouse interleukin-1 beta (IL-1beta) in various biological samples It uses electrochemiluminescence (ECL) detection offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 384-well plate with capture antibodies ECL-tagged detection antibodies calibrators and buffers for 384 tests It quantifies IL-1beta from 50-500 000 pg/mL with 10% CV precision The assay completes in 3 5 hours
Catalog No. 50-000-79339 Supplier Meso Scale Discovery Supplier No. K252TUK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-1beta 384-well Assay Kit is a multiplex immunoassay for measuring mouse interleukin-1 beta (IL-1beta) in various biological samples It uses electrochemiluminescence (ECL) detection offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 384-well plate with capture antibodies ECL-tagged detection antibodies calibrators and buffers for 384 tests It quantifies IL-1beta from 50-500 000 pg/mL with 10% CV precision The assay completes in 3 5 hours