Learn More
Meso Scale Discovery UPLEX MSE IL2 ASY384well
SDP
Catalog No. 5000081218
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-2 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 2 (IL-2) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and buffers for 384 tests It can detect mouse IL-2 with a lower limit of detection of 0 06 pg/mL and an upper limit of 100 000 pg/mL The assay takes 3 5 hours to complete
Catalog No. 50-000-81218 Supplier Meso Scale Discovery Supplier No. K252TVK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-2 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 2 (IL-2) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and buffers for 384 tests It can detect mouse IL-2 with a lower limit of detection of 0 06 pg/mL and an upper limit of 100 000 pg/mL The assay takes 3 5 hours to complete