Learn More
Meso Scale Discovery UPLEX MSE IL21 ASY384well
SDP
Catalog No. 5000081284
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-21 384-Well Assay Kit is designed for the quantitative measurement of mouse interleukin 21 (IL-21) in serum plasma cell culture supernatants cell and tissue lysates using an electrochemiluminescent detection technology The assay combines capture antibodies on distinct spots within each well with detection antibodies labeled with electrochemiluminescent labels (MSD GOLD) The kit includes a 384-well plate coated with capture antibodies detection antibodies calibrator diluent wash buffer read buffer 10X mouse IL-21 calibrator and a plate seal The assay has a typical sensitivity of 0 3 pg/mL and an assay range from 0 4 - 2500 pg/mL
Catalog No. 50-000-81284 Supplier Meso Scale Discovery Supplier No. K252WBK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-21 384-Well Assay Kit is designed for the quantitative measurement of mouse interleukin 21 (IL-21) in serum plasma cell culture supernatants cell and tissue lysates using an electrochemiluminescent detection technology The assay combines capture antibodies on distinct spots within each well with detection antibodies labeled with electrochemiluminescent labels (MSD GOLD) The kit includes a 384-well plate coated with capture antibodies detection antibodies calibrator diluent wash buffer read buffer 10X mouse IL-21 calibrator and a plate seal The assay has a typical sensitivity of 0 3 pg/mL and an assay range from 0 4 - 2500 pg/mL