Learn More
Meso Scale Discovery UPLEX MSE IL22 ASY384well
SDP
Catalog No. 5000079322
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-22 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 22 (IL-22) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and buffers for 384 tests It can accurately quantify IL-22 concentrations and is suitable for mouse cytokine profiling and immunology studies
Catalog No. 50-000-79322 Supplier Meso Scale Discovery Supplier No. K252WIK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-22 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 22 (IL-22) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and buffers for 384 tests It can accurately quantify IL-22 concentrations and is suitable for mouse cytokine profiling and immunology studies