Learn More
Meso Scale Discovery UPLEX MSE IL23 ASY384well
SDP
Catalog No. 5000081205
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-23 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 23 (IL-23) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and controls The assay takes approximately 3 5 hours to complete and requires a small sample volume of 25 L This product allows for the accurate and reliable quantification of IL-23
Catalog No. 50-000-81205 Supplier Meso Scale Discovery Supplier No. K252WGK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-23 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 23 (IL-23) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and controls The assay takes approximately 3 5 hours to complete and requires a small sample volume of 25 L This product allows for the accurate and reliable quantification of IL-23