Learn More
Meso Scale Discovery UPLEX MSE IL27p28/IL30 ASY
SDP
Catalog No. 5000082110
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-27p28/IL-30 Assay Kit is a multiplex immunoassay that measures mouse IL-27p28 and IL-30 in serum plasma and cell culture supernatants This assay uses MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent Results can be read on MSD s SECTOR Imager and analyzed with MSD Discovery Workbench software This assay allows for the simultaneous quantitative measurement of multiple analytes from a single sample
Catalog No. 50-000-82110 Supplier Meso Scale Discovery Supplier No. K152WCK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-27p28/IL-30 Assay Kit is a multiplex immunoassay that measures mouse IL-27p28 and IL-30 in serum plasma and cell culture supernatants This assay uses MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent Results can be read on MSD s SECTOR Imager and analyzed with MSD Discovery Workbench software This assay allows for the simultaneous quantitative measurement of multiple analytes from a single sample