Learn More
Meso Scale Discovery UPLEX MSE IL27p28/IL30 ASY384
SDP
Catalog No. 5000079976
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-27p28/IL-30 384-Well Assay Kit is a multiplex immunoassay for measuring mouse IL-27p28 and IL-30 in serum plasma and cell culture supernatants It uses electrochemiluminescence detection in a 384-well plate enabling high-throughput analysis of up to 80 samples in duplicate With a sensitivity of 0 3-2 4 pg/mL it requires 12 5 L of sample The kit includes a plate with capture antibodies detection antibodies with an electrochemiluminescent label and calibrator solutions for standard curves measuring natural and recombinant mouse IL-27p28 and IL-30
Catalog No. 50-000-79976 Supplier Meso Scale Discovery Supplier No. K252WCK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-27p28/IL-30 384-Well Assay Kit is a multiplex immunoassay for measuring mouse IL-27p28 and IL-30 in serum plasma and cell culture supernatants It uses electrochemiluminescence detection in a 384-well plate enabling high-throughput analysis of up to 80 samples in duplicate With a sensitivity of 0 3-2 4 pg/mL it requires 12 5 L of sample The kit includes a plate with capture antibodies detection antibodies with an electrochemiluminescent label and calibrator solutions for standard curves measuring natural and recombinant mouse IL-27p28 and IL-30