Learn More
Meso Scale Discovery UPLEX MSE IL31 ASY384well
SDP
Catalog No. 5000079974
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-31 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 31 (IL-31) in serum plasma and cell culture supernatants The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible detection of mouse IL-31 with a lower limit of detection of 0 03 pg/mL and an upper limit of quantification of 2500 pg/mL The kit includes a mouse IL-31 calibrator and positive control detection antibody read buffer 10X wash buffer blocker A and 384-well plates coated with capture antibody Sample incubation time is 2 hours
Catalog No. 50-000-79974 Supplier Meso Scale Discovery Supplier No. K252WEK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-31 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 31 (IL-31) in serum plasma and cell culture supernatants The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible detection of mouse IL-31 with a lower limit of detection of 0 03 pg/mL and an upper limit of quantification of 2500 pg/mL The kit includes a mouse IL-31 calibrator and positive control detection antibody read buffer 10X wash buffer blocker A and 384-well plates coated with capture antibody Sample incubation time is 2 hours