Learn More
Meso Scale Discovery UPLEX MSE IL33 ASY384well
SDP
Catalog No. 5000079975
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-33 384-Well Assay Kit is an electrochemiluminescent immunoassay for quantifying mouse interleukin 33 (IL-33) in various samples It uses MSD s SULFO-TAG labels and multi-array technology for a rapid sensitive and accurate assay with a dynamic range of up to 4 logs The kit includes a 384-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and diluent Sensitivity is 0 3 pg/mL with an upper quantitation limit of 10 000 pg/mL
Catalog No. 50-000-79975 Supplier Meso Scale Discovery Supplier No. K252WFK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-33 384-Well Assay Kit is an electrochemiluminescent immunoassay for quantifying mouse interleukin 33 (IL-33) in various samples It uses MSD s SULFO-TAG labels and multi-array technology for a rapid sensitive and accurate assay with a dynamic range of up to 4 logs The kit includes a 384-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and diluent Sensitivity is 0 3 pg/mL with an upper quantitation limit of 10 000 pg/mL