Learn More
Meso Scale Discovery UPLEX MSE IL5 ASY384well
SDP
Catalog No. 5000079324
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-5 Assay Kit is a highly sensitive immunoassay for the quantitative measurement of mouse interleukin 5 (IL-5) in serum plasma and cell culture supernatants The assay uses MSD s proprietary electrochemiluminescent detection technology and provides exceptional sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibody detection antibody read buffer calibrator diluent wash buffer and sample diluent The assay takes approximately 3 5 hours to complete and can detect mouse IL-5 concentrations as low as 0 08 pg/mL
Catalog No. 50-000-79324 Supplier Meso Scale Discovery Supplier No. K252UOK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-5 Assay Kit is a highly sensitive immunoassay for the quantitative measurement of mouse interleukin 5 (IL-5) in serum plasma and cell culture supernatants The assay uses MSD s proprietary electrochemiluminescent detection technology and provides exceptional sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibody detection antibody read buffer calibrator diluent wash buffer and sample diluent The assay takes approximately 3 5 hours to complete and can detect mouse IL-5 concentrations as low as 0 08 pg/mL