Learn More
Meso Scale Discovery UPLEX MSE IL6 ASY384well
SDP
Catalog No. 5000079979
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-6 384-Well Assay Kit is an electrochemiluminescent immunoassay for quantifying mouse interleukin 6 (IL-6) in serum plasma tissue culture supernatant and other samples This kit uses MSD s SULFO-TAG labels and multi-array technology for sensitive and accurate IL-6 measurement with a broad dynamic range It has high specificity and no cross-reactivity with other cytokines Each kit includes reagents for one 384-well plate such as capture antibody-coated plates detection antibodies calibrators controls buffers and substrate The assay takes about 4 hours
Catalog No. 50-000-79979 Supplier Meso Scale Discovery Supplier No. K252TXK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-6 384-Well Assay Kit is an electrochemiluminescent immunoassay for quantifying mouse interleukin 6 (IL-6) in serum plasma tissue culture supernatant and other samples This kit uses MSD s SULFO-TAG labels and multi-array technology for sensitive and accurate IL-6 measurement with a broad dynamic range It has high specificity and no cross-reactivity with other cytokines Each kit includes reagents for one 384-well plate such as capture antibody-coated plates detection antibodies calibrators controls buffers and substrate The assay takes about 4 hours