Learn More
Meso Scale Discovery UPLEX MSE IL9 ASY384well
SDP
Catalog No. 5000081316
Encompass
The Meso Scale Discovery U-PLEX Mouse IL-9 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 9 (IL-9) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and buffers It allows simultaneous measurement of IL-9 along with other biomarkers The assay takes 3 5 hours to complete and only requires 10 L of sample per well
Catalog No. 50-000-81316 Supplier Meso Scale Discovery Supplier No. K252XKK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IL-9 384-Well Assay Kit is a multiplex immunoassay that measures mouse interleukin 9 (IL-9) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and buffers It allows simultaneous measurement of IL-9 along with other biomarkers The assay takes 3 5 hours to complete and only requires 10 L of sample per well