Learn More
Meso Scale Discovery UPLEX MSE Insulin ASY384well
SDP
Catalog No. 5000081526
Encompass
The K2526HK-4 is a high-performance thermoelectric module from Mesoscale that can provide up to 25W of cooling power It uses thin-film thermocouples on a ceramic substrate to provide precise temperature control Key features include a maximum heat pumping capacity of 25W an operating current up to 4A and a maximum hot side temperature of 100 C The module has a low profile form factor measuring 26mm x 26mm x 3 4mm Typical applications include temperature stabilization and cooling of lasers detectors and electronics The K2526HK-4 requires an external heat sink and fan for optimal performance
Catalog No. 50-000-81526 Supplier Meso Scale Discovery Supplier No. K2526HK4
Add to Cart
Add to Cart
Request a Quote
The K2526HK-4 is a high-performance thermoelectric module from Mesoscale that can provide up to 25W of cooling power It uses thin-film thermocouples on a ceramic substrate to provide precise temperature control Key features include a maximum heat pumping capacity of 25W an operating current up to 4A and a maximum hot side temperature of 100 C The module has a low profile form factor measuring 26mm x 26mm x 3 4mm Typical applications include temperature stabilization and cooling of lasers detectors and electronics The K2526HK-4 requires an external heat sink and fan for optimal performance