Learn More
Meso Scale Discovery UPLEX MSE IP10 ASY384well
SDP
Catalog No. 5000078967
Encompass
The Meso Scale Discovery U-PLEX Mouse IP-10 (384-Well Assay Kit) is an electrochemiluminescent immunoassay for quantifying IP-10 in mouse serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive accurate results in 3 5 hours The kit includes a 384-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and a read buffer for 384 tests It has a dynamic range of 1 95 to 10 000 pg/mL and requires 25 L per well offering high-throughput measurement with high sensitivity and low sample use
Catalog No. 50-000-78967 Supplier Meso Scale Discovery Supplier No. K252UFK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IP-10 (384-Well Assay Kit) is an electrochemiluminescent immunoassay for quantifying IP-10 in mouse serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive accurate results in 3 5 hours The kit includes a 384-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and a read buffer for 384 tests It has a dynamic range of 1 95 to 10 000 pg/mL and requires 25 L per well offering high-throughput measurement with high sensitivity and low sample use