Learn More
Meso Scale Discovery UPLEX MSE KC/GRO ASY384well
SDP
Catalog No. 5000081282
Encompass
The Meso Scale Discovery U-PLEX Mouse KC/GRO/CXCL1 Kit is a multiplex immunoassay that measures mouse Keratinocyte Chemoattractant (KC/GRO/CXCL1) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and assay diluent The assay takes approximately 3 5 hours to run and 10 L of sample is required
Catalog No. 50-000-81282 Supplier Meso Scale Discovery Supplier No. K252VWK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse KC/GRO/CXCL1 Kit is a multiplex immunoassay that measures mouse Keratinocyte Chemoattractant (KC/GRO/CXCL1) in serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and assay diluent The assay takes approximately 3 5 hours to run and 10 L of sample is required