Learn More
Meso Scale Discovery UPLEX MSE Leptin ASYQP
SDP
Catalog No. 5000082173
Encompass
The Meso Scale Discovery U-PLEX Mouse Leptin Assay Kit is a multiplex immunoassay that measures mouse leptin in serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls The assay has a lower limit of detection of 1 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for leptin The kit can perform up to 80 tests in duplicate and results are obtained in 3 5 hours
Catalog No. 50-000-82173 Supplier Meso Scale Discovery Supplier No. K1525ZK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse Leptin Assay Kit is a multiplex immunoassay that measures mouse leptin in serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls The assay has a lower limit of detection of 1 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for leptin The kit can perform up to 80 tests in duplicate and results are obtained in 3 5 hours