Learn More
Meso Scale Discovery UPLEX MSE MCP1 ASY384well
SDP
Catalog No. 5000079320
Encompass
The Meso Scale Discovery U-PLEX Mouse MCP-1 384-well Assay Kit is a multiplex immunoassay that measures mouse monocyte chemoattractant protein 1 (MCP-1) in serum plasma and cell culture supernatants The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision It requires just 25 L of sample per well and the 384-well plate allows high-throughput analysis The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-79320 Supplier Meso Scale Discovery Supplier No. K252UGK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse MCP-1 384-well Assay Kit is a multiplex immunoassay that measures mouse monocyte chemoattractant protein 1 (MCP-1) in serum plasma and cell culture supernatants The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision It requires just 25 L of sample per well and the 384-well plate allows high-throughput analysis The assay takes approximately 3 5 hours to complete