Learn More
Meso Scale Discovery UPLEX MSE MCP5/CCL12 ASY
SDP
Catalog No. 5000082003
Encompass
The Meso Scale Discovery U-PLEX Mouse MCP-5 (CCL12) Assay is a multiplex immunoassay kit that enables simultaneous quantification of mouse MCP-5 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can perform up to 70 tests across two plates The lower limit of detection for mouse MCP-5 is 1 3 pg/mL
Catalog No. 50-000-82003 Supplier Meso Scale Discovery Supplier No. K152Y1K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse MCP-5 (CCL12) Assay is a multiplex immunoassay kit that enables simultaneous quantification of mouse MCP-5 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can perform up to 70 tests across two plates The lower limit of detection for mouse MCP-5 is 1 3 pg/mL