Learn More
Meso Scale Discovery UPLEX MSE MCP5/CCL12 ASY384
SDP
Catalog No. 5000079467
Encompass
The Meso Scale Discovery U-PLEX Mouse MCP-5 (CCL12) 384-well Assay Kit is designed for the quantitative measurement of mouse MCP-5 in serum plasma cell culture supernatants cell and tissue lysates and other biological samples This kit uses electrochemiluminescence (ECL) technology and provides high sensitivity broad dynamic range and rapid time to results The kit includes a 384-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls The assay can detect mouse MCP-5 in the range of 2 4-10 000 pg/mL
Catalog No. 50-000-79467 Supplier Meso Scale Discovery Supplier No. K252Y1K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse MCP-5 (CCL12) 384-well Assay Kit is designed for the quantitative measurement of mouse MCP-5 in serum plasma cell culture supernatants cell and tissue lysates and other biological samples This kit uses electrochemiluminescence (ECL) technology and provides high sensitivity broad dynamic range and rapid time to results The kit includes a 384-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls The assay can detect mouse MCP-5 in the range of 2 4-10 000 pg/mL