Learn More
Meso Scale Discovery UPLEX MSE MCP5/CCL12 ASY384
SDP
Catalog No. 5000081318
Encompass
The Meso Scale Discovery U-PLEX Mouse MCP-5 (CCL12) 384-well Assay Kit is designed to quantitatively measure mouse MCP-5 (CCL12) in serum plasma cell culture supernatants cell and tissue lysates using an electrochemiluminescent detection method The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and buffer for 384 tests The assay has a typical sensitivity of 0 3 pg/mL and a 7-log dynamic range from 0 3-10 000 pg/mL This immunoassay allows high-throughput sensitive and accurate quantification of mouse MCP-5 (CCL12)
Catalog No. 50-000-81318 Supplier Meso Scale Discovery Supplier No. K252Y1K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse MCP-5 (CCL12) 384-well Assay Kit is designed to quantitatively measure mouse MCP-5 (CCL12) in serum plasma cell culture supernatants cell and tissue lysates using an electrochemiluminescent detection method The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and buffer for 384 tests The assay has a typical sensitivity of 0 3 pg/mL and a 7-log dynamic range from 0 3-10 000 pg/mL This immunoassay allows high-throughput sensitive and accurate quantification of mouse MCP-5 (CCL12)