Learn More
Meso Scale Discovery UPLEX MSE MIP1 ASYQP
SDP
Catalog No. 5000079336
Encompass
The U-PLEX Gateway assay kits from Meso Scale Discovery enable researchers to create custom multiplex immunoassays for simultaneous measurement of multiple biomarkers in a single sample These electrochemiluminescence-based assays offer superior sensitivity wide dynamic range and reduced sample volume requirements compared to traditional methods The platform supports flexible assay development with pre-validated antibody pairs and allows detection of up to 10 analytes per well making it ideal for biomarker discovery drug development and clinical research applications
Catalog No. 50-000-79336 Supplier Meso Scale Discovery Supplier No. K152UJK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Gateway assay kits from Meso Scale Discovery enable researchers to create custom multiplex immunoassays for simultaneous measurement of multiple biomarkers in a single sample These electrochemiluminescence-based assays offer superior sensitivity wide dynamic range and reduced sample volume requirements compared to traditional methods The platform supports flexible assay development with pre-validated antibody pairs and allows detection of up to 10 analytes per well making it ideal for biomarker discovery drug development and clinical research applications