Learn More
Meso Scale Discovery UPLEX MSE MIP3 ASY384well
SDP
Catalog No. 5000081281
Encompass
The Meso Scale Discovery U-PLEX Mouse MIP-3alpha 384-Well Assay Kit is a multiplex immunoassay that measures mouse macrophage inflammatory protein 3 alpha (MIP-3alpha) in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent label and calibrator It allows simultaneous detection of mouse MIP-3alpha along with other biomarkers in the sample for understanding immune response
Catalog No. 50-000-81281 Supplier Meso Scale Discovery Supplier No. K252UZK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse MIP-3alpha 384-Well Assay Kit is a multiplex immunoassay that measures mouse macrophage inflammatory protein 3 alpha (MIP-3alpha) in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent label and calibrator It allows simultaneous detection of mouse MIP-3alpha along with other biomarkers in the sample for understanding immune response