Learn More
Meso Scale Discovery UPLEX MSE MMP9 TTL ASY
SDP
Catalog No. 5000082000
Encompass
The Meso Scale Discovery U-PLEX Mouse MMP-9 Total Assay Kit is a multiplex immunoassay that measures total MMP-9 levels in mouse biological samples such as serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator and controls The assay has a typical sensitivity of 0 04 ng/mL and an upper limit of quantification of 2500 ng/mL for MMP-9 It requires a small sample volume of 25 L and the total assay time is approximately 3 5 hours
Catalog No. 50-000-82000 Supplier Meso Scale Discovery Supplier No. K152ZGK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse MMP-9 Total Assay Kit is a multiplex immunoassay that measures total MMP-9 levels in mouse biological samples such as serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator and controls The assay has a typical sensitivity of 0 04 ng/mL and an upper limit of quantification of 2500 ng/mL for MMP-9 It requires a small sample volume of 25 L and the total assay time is approximately 3 5 hours