Learn More
Meso Scale Discovery UPLEX MSE MMP9 TTL ASY384well
SDP
Catalog No. 5000080204
Encompass
The Meso Scale Discovery Mouse MMP-9 Total Kit is a multiplex immunoassay that quantitatively measures the total amount of mouse matrix metalloproteinase 9 (MMP-9) in serum plasma and cell culture supernatants The assay uses MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer 10X Diluent and calibrator The total assay time is 3 5 hours This product can be used to study MMP-9 levels in mouse models of disease
Catalog No. 50-000-80204 Supplier Meso Scale Discovery Supplier No. K252ZGK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Mouse MMP-9 Total Kit is a multiplex immunoassay that quantitatively measures the total amount of mouse matrix metalloproteinase 9 (MMP-9) in serum plasma and cell culture supernatants The assay uses MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer 10X Diluent and calibrator The total assay time is 3 5 hours This product can be used to study MMP-9 levels in mouse models of disease