Learn More
Meso Scale Discovery UPLEX MSE MMP9 TTL ASYQP
SDP
Catalog No. 5000079113
Encompass
The Meso Scale Discovery U-PLEX Mouse MMP-9 Total Assay Kit measures total MMP-9 in mouse samples like serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection for high sensitivity and a broad dynamic range with low sample volume The kit includes a 96-well plate with capture and detection antibodies Read Buffer 10X Diluent and calibrator It detects MMP-9 levels from 0 03-1500 ng/mL with high specificity and minimal cross-reactivity enabling rapid and reliable quantification of total MMP-9
Catalog No. 50-000-79113 Supplier Meso Scale Discovery Supplier No. K152ZGK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse MMP-9 Total Assay Kit measures total MMP-9 in mouse samples like serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection for high sensitivity and a broad dynamic range with low sample volume The kit includes a 96-well plate with capture and detection antibodies Read Buffer 10X Diluent and calibrator It detects MMP-9 levels from 0 03-1500 ng/mL with high specificity and minimal cross-reactivity enabling rapid and reliable quantification of total MMP-9