Learn More
Meso Scale Discovery UPLEX MSE NGAL/LCN2 ASY
SDP
Catalog No. 5000082005
Encompass
The Meso Scale Discovery U-PLEX Mouse NGAL (LCN2) Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Neutrophil Gelatinase-Associated Lipocalin (NGAL) in mouse biological samples such as serum plasma urine and cell culture supernatants This assay uses MSD s proprietary U-PLEX technology is highly sensitive has a broad dynamic range of 78 - 50 000 pg/mL and shows no cross-reactivity with other mouse proteins The assay requires a small sample volume of 50 L and provides results in under 3 hours This product can aid research on kidney injury and other inflammatory conditions involving NGAL
Catalog No. 50-000-82005 Supplier Meso Scale Discovery Supplier No. K152Z1K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse NGAL (LCN2) Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Neutrophil Gelatinase-Associated Lipocalin (NGAL) in mouse biological samples such as serum plasma urine and cell culture supernatants This assay uses MSD s proprietary U-PLEX technology is highly sensitive has a broad dynamic range of 78 - 50 000 pg/mL and shows no cross-reactivity with other mouse proteins The assay requires a small sample volume of 50 L and provides results in under 3 hours This product can aid research on kidney injury and other inflammatory conditions involving NGAL