Learn More
Meso Scale Discovery UPLEX MSE NGAL/LCN2 ASY384
SDP
Catalog No. 5000080011
Encompass
The Meso Scale Discovery U-PLEX Mouse NGAL (LCN2) 384-well assay is a multiplex immunoassay panel that measures natural and recombinant mouse Lipocalin-2/NGAL in serum plasma urine cell culture supernatants cell and tissue lysates and other biological samples This assay utilizes MSD s electrochemiluminescent detection technology and is provided in a 384-well plate format allowing high-throughput analysis of up to 80 samples for mouse NGAL The sensitivity of the assay is excellent with a lower limit of detection of 98 pg/mL This quantitative immunoassay provides accurate reproducible data for measuring natural and recombinant mouse NGAL with a broad dynamic range
Catalog No. 50-000-80011 Supplier Meso Scale Discovery Supplier No. K252Z1K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse NGAL (LCN2) 384-well assay is a multiplex immunoassay panel that measures natural and recombinant mouse Lipocalin-2/NGAL in serum plasma urine cell culture supernatants cell and tissue lysates and other biological samples This assay utilizes MSD s electrochemiluminescent detection technology and is provided in a 384-well plate format allowing high-throughput analysis of up to 80 samples for mouse NGAL The sensitivity of the assay is excellent with a lower limit of detection of 98 pg/mL This quantitative immunoassay provides accurate reproducible data for measuring natural and recombinant mouse NGAL with a broad dynamic range