Learn More
Meso Scale Discovery UPLEX MSE NGAL/LCN2 ASY384well
SDP
Catalog No. 5000080200
Encompass
The Meso Scale Discovery U-PLEX Mouse NGAL (LCN2) 384-well assay is a multiplex immunoassay panel that measures natural and recombinant mouse Lipocalin-2/NGAL in serum plasma urine cell culture supernatants cell and tissue lysates and other biological samples This assay uses MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements It is available as a 384-well plate that includes standards controls calibrators and reagents for the quantitative analysis of natural and recombinant mouse Lipocalin-2/NGAL
Catalog No. 50-000-80200 Supplier Meso Scale Discovery Supplier No. K252Z1K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse NGAL (LCN2) 384-well assay is a multiplex immunoassay panel that measures natural and recombinant mouse Lipocalin-2/NGAL in serum plasma urine cell culture supernatants cell and tissue lysates and other biological samples This assay uses MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements It is available as a 384-well plate that includes standards controls calibrators and reagents for the quantitative analysis of natural and recombinant mouse Lipocalin-2/NGAL