Learn More
Meso Scale Discovery UPLEX MSE PYY TTL ASY
SDP
Catalog No. 5000082074
Encompass
The Meso Scale Discovery U-PLEX Mouse PYY Total Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of total Peptide YY (PYY) in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements This kit includes a 96-well plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and sample diluent The assay has a typical sensitivity of 1 5 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-82074 Supplier Meso Scale Discovery Supplier No. K1526BK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse PYY Total Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of total Peptide YY (PYY) in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements This kit includes a 96-well plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and sample diluent The assay has a typical sensitivity of 1 5 pg/mL and an upper limit of quantification of 10 000 pg/mL