Learn More
Meso Scale Discovery UPLEX MSE PYY TTL ASY384well
SDP
Catalog No. 5000079983
Encompass
The Meso Scale Discovery U-PLEX Mouse PYY Total 384-Well Assay Kit is designed to measure total PYY levels in mouse serum plasma and cell culture supernatants This electrochemiluminescence immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and buffers for 384 tests The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL This product enables high-throughput quantification of total PYY
Catalog No. 50-000-79983 Supplier Meso Scale Discovery Supplier No. K2526BK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse PYY Total 384-Well Assay Kit is designed to measure total PYY levels in mouse serum plasma and cell culture supernatants This electrochemiluminescence immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and buffers for 384 tests The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL This product enables high-throughput quantification of total PYY