Learn More
Meso Scale Discovery UPLEX MSE RANTES ASY384well
SDP
Catalog No. 5000078968
Encompass
The Meso Scale Discovery RANTES Kit is a sensitive electrochemiluminescent immunoassay for measuring RANTES in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology detecting RANTES from 0 64 to 10 000 pg/mL The assay is specific for human RANTES with no cross-reactivity The kit includes a 96-well plate detection antibody calibrators controls and instructions Results are available in 3 5 hours using an MSD instrument
Catalog No. 50-000-78968 Supplier Meso Scale Discovery Supplier No. K252A2K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery RANTES Kit is a sensitive electrochemiluminescent immunoassay for measuring RANTES in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology detecting RANTES from 0 64 to 10 000 pg/mL The assay is specific for human RANTES with no cross-reactivity The kit includes a 96-well plate detection antibody calibrators controls and instructions Results are available in 3 5 hours using an MSD instrument