Learn More
Meso Scale Discovery UPLEX MSE SDF1 ASY384well
SDP
Catalog No. 5000079440
Encompass
The U-PLEX Mouse SDF-1alpha 384-well Assay Kit from Meso Scale Discovery is a 384-well immunoassay that quantitatively measures mouse SDF-1alpha (CXCL12) in serum plasma cell culture supernatants cell and tissue lysates This assay uses electrochemiluminescence (ECL) detection and provides high sensitivity broad dynamic range and low sample volume requirements The kit includes a 384-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and positive control The assay can detect mouse SDF-1alpha with a lower limit of detection of 1 5 pg/mL and an upper limit of 100 000 pg/mL
Catalog No. 50-000-79440 Supplier Meso Scale Discovery Supplier No. K252VBK4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Mouse SDF-1alpha 384-well Assay Kit from Meso Scale Discovery is a 384-well immunoassay that quantitatively measures mouse SDF-1alpha (CXCL12) in serum plasma cell culture supernatants cell and tissue lysates This assay uses electrochemiluminescence (ECL) detection and provides high sensitivity broad dynamic range and low sample volume requirements The kit includes a 384-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and positive control The assay can detect mouse SDF-1alpha with a lower limit of detection of 1 5 pg/mL and an upper limit of 100 000 pg/mL