Learn More
Meso Scale Discovery UPLEX MSE SDF1 ASYQP
SDP
Catalog No. 5000078717
Encompass
The Meso Scale Discovery U-PLEX Mouse SDF-1alpha Assay is a multiplex immunoassay kit that enables the simultaneous quantitative measurement of mouse SDF-1alpha in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with a rapid 2 5 hour turnaround time from sample to results This assay requires a Meso Scale Discovery instrument and provides components for one 96-well plate testing
Catalog No. 50-000-78717 Supplier Meso Scale Discovery Supplier No. K152VBK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse SDF-1alpha Assay is a multiplex immunoassay kit that enables the simultaneous quantitative measurement of mouse SDF-1alpha in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with a rapid 2 5 hour turnaround time from sample to results This assay requires a Meso Scale Discovery instrument and provides components for one 96-well plate testing