Learn More
Meso Scale Discovery UPLEX MSE TARC ASY384well
SDP
Catalog No. 5000081528
Encompass
The Meso Scale Discovery U-PLEX Mouse TARC 384-Well Assay Kit is a multiplex immunoassay that measures TARC (thymus and activation regulated chemokine) in mouse serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides a 384-well plate format with a testing range of 0 03-2500 pg/mL This assay allows for the quantitative measurement of TARC which plays a role in inflammatory responses It requires a low sample volume of 25 L and the total assay time is approximately 3 5 hours This kit includes a 384-well standard microplate detection antibody read buffer 10X diluent and vials of calibrator
Catalog No. 50-000-81528 Supplier Meso Scale Discovery Supplier No. K252B0K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse TARC 384-Well Assay Kit is a multiplex immunoassay that measures TARC (thymus and activation regulated chemokine) in mouse serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides a 384-well plate format with a testing range of 0 03-2500 pg/mL This assay allows for the quantitative measurement of TARC which plays a role in inflammatory responses It requires a low sample volume of 25 L and the total assay time is approximately 3 5 hours This kit includes a 384-well standard microplate detection antibody read buffer 10X diluent and vials of calibrator