Learn More
Meso Scale Discovery UPLEX MSE TGFB1 ASYQP
SDP
Catalog No. 5000078696
Encompass
The Meso Scale Discovery U-PLEX Mouse TGF-beta1 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of TGF-beta1 in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides a sensitive and rapid method for measuring TGF-beta1 with a detection range of 1 7-1500 pg/mL and an LLOQ of 1 7 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody calibrator diluent wash buffer read buffer and a vial of lyophilized calibrator The total assay time is 3 5 hours This assay allows high-throughput quantitative analysis of TGF-beta1 from mouse samples
Catalog No. 50-000-78696 Supplier Meso Scale Discovery Supplier No. K152XWK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse TGF-beta1 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of TGF-beta1 in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides a sensitive and rapid method for measuring TGF-beta1 with a detection range of 1 7-1500 pg/mL and an LLOQ of 1 7 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody calibrator diluent wash buffer read buffer and a vial of lyophilized calibrator The total assay time is 3 5 hours This assay allows high-throughput quantitative analysis of TGF-beta1 from mouse samples