Learn More
Meso Scale Discovery UPLEX MSE TGFB3 ASY384well
SDP
Catalog No. 5000081190
Encompass
The Meso Scale Discovery U-PLEX Mouse TGF-beta3 384-well Assay Kit is a multiplex immunoassay that measures TGF-beta3 levels in mouse serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This 384-well assay allows high throughput screening and only requires 10-25 L sample volume per well The kit includes a plate coated with capture antibodies detection antibodies Read Buffer and calibrator It can detect TGF-beta3 concentrations as low as 17 pg/mL with an upper limit of 50 000 pg/mL
Catalog No. 50-000-81190 Supplier Meso Scale Discovery Supplier No. K252XVK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse TGF-beta3 384-well Assay Kit is a multiplex immunoassay that measures TGF-beta3 levels in mouse serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This 384-well assay allows high throughput screening and only requires 10-25 L sample volume per well The kit includes a plate coated with capture antibodies detection antibodies Read Buffer and calibrator It can detect TGF-beta3 concentrations as low as 17 pg/mL with an upper limit of 50 000 pg/mL