Learn More
Meso Scale Discovery UPLEX MSE TGFB3 ASYQP
SDP
Catalog No. 5000079111
Encompass
The Meso Scale Discovery U-PLEX Mouse TGF-beta3 Assay Kit measures TGF-beta3 in mouse samples like serum plasma and tissue lysates This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels for sensitive quantification down to sub-pg/mL It includes a 96-well plate with capture antibodies detection antibodies calibrators and diluent The assay takes about 3 5 hours and suits lab automation It enables high-throughput analysis of TGF-beta3 a key cytokine in development immune regulation cancer and fibrosis
Catalog No. 50-000-79111 Supplier Meso Scale Discovery Supplier No. K152XVK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse TGF-beta3 Assay Kit measures TGF-beta3 in mouse samples like serum plasma and tissue lysates This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels for sensitive quantification down to sub-pg/mL It includes a 96-well plate with capture antibodies detection antibodies calibrators and diluent The assay takes about 3 5 hours and suits lab automation It enables high-throughput analysis of TGF-beta3 a key cytokine in development immune regulation cancer and fibrosis