Learn More
Meso Scale Discovery UPLEX MSE TNFRI ASY384well
SDP
Catalog No. 5000081040
Encompass
The Meso Scale Discovery U-PLEX Mouse TNF RI 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of mouse tumor necrosis factor receptor 1 (TNFR1) in serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results in just 3 5 hours The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and buffer The assay has a sensitivity of 0 3 pg/mL and a broad dynamic range from 0 3-10 000 pg/mL
Catalog No. 50-000-81040 Supplier Meso Scale Discovery Supplier No. K2520VK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse TNF RI 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of mouse tumor necrosis factor receptor 1 (TNFR1) in serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results in just 3 5 hours The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrator and buffer The assay has a sensitivity of 0 3 pg/mL and a broad dynamic range from 0 3-10 000 pg/mL