Learn More
Meso Scale Discovery UPLEX MSE TNFRI ASYQP
SDP
Catalog No. 5000079388
Encompass
The Meso Scale Discovery U-PLEX Mouse TNF RI Assay is a multiplex immunoassay kit for measuring mouse TNFR1 in serum plasma and cell culture supernatants This ECL-based assay uses MSD s SULFO-TAG labels and multi-array technology for sensitive results from small samples The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are analyzed on MSD s SECTOR Imager offering a rapid method to quantify soluble TNFR1 levels for inflammation and autoimmunity research
Catalog No. 50-000-79388 Supplier Meso Scale Discovery Supplier No. K1520VK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse TNF RI Assay is a multiplex immunoassay kit for measuring mouse TNFR1 in serum plasma and cell culture supernatants This ECL-based assay uses MSD s SULFO-TAG labels and multi-array technology for sensitive results from small samples The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are analyzed on MSD s SECTOR Imager offering a rapid method to quantify soluble TNFR1 levels for inflammation and autoimmunity research