Learn More
Meso Scale Discovery UPLEX MSE VEGFa ASY384well
SDP
Catalog No. 5000081260
Encompass
The Meso Scale Discovery U-PLEX Mouse VEGF-A 384-Well Assay Kit is a multiplex immunoassay that measures VEGF-A levels in mouse serum plasma and cell culture supernatants The kit uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision It includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound calibrators and controls The assay has a lower limit of detection of 0 61 pg/mL and an upper limit of quantification of 10 000 pg/mL for VEGF-A It requires a small sample volume of 25 L and the total assay time is approximately 3 5 hours
Catalog No. 50-000-81260 Supplier Meso Scale Discovery Supplier No. K252UVK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse VEGF-A 384-Well Assay Kit is a multiplex immunoassay that measures VEGF-A levels in mouse serum plasma and cell culture supernatants The kit uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision It includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound calibrators and controls The assay has a lower limit of detection of 0 61 pg/mL and an upper limit of quantification of 10 000 pg/mL for VEGF-A It requires a small sample volume of 25 L and the total assay time is approximately 3 5 hours