Learn More
Meso Scale Discovery UPLEX NHP ENA78 ASYQP 25 PL
SDP
Catalog No. 5000080296
Encompass
The U-PLEX NHP ENA-78 Assay (K156VEK) is a multiplex immunoassay kit from Meso Scale Discovery designed for quantitative measurement of ENA-78 (CXCL5) protein levels in non-human primate samples This electrochemiluminescence-based assay provides high sensitivity detection with minimal sample volume requirements featuring pre-coated plates and calibrated standards for reliable reproducible results in research applications studying inflammatory responses and chemokine signaling pathways
Catalog No. 50-000-80296 Supplier Meso Scale Discovery Supplier No. K156VEK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP ENA-78 Assay (K156VEK) is a multiplex immunoassay kit from Meso Scale Discovery designed for quantitative measurement of ENA-78 (CXCL5) protein levels in non-human primate samples This electrochemiluminescence-based assay provides high sensitivity detection with minimal sample volume requirements featuring pre-coated plates and calibrated standards for reliable reproducible results in research applications studying inflammatory responses and chemokine signaling pathways