Learn More
Meso Scale Discovery UPLEX NHP Eotaxin2 ASY 25 PL
SDP
Catalog No. 5000078372
Encompass
The Meso Scale Discovery U-PLEX NHP Eotaxin-2 Assay is a multiplex immunoassay kit for measuring Eotaxin-2 in non-human primate samples like serum and plasma It uses MSD s electrochemiluminescent detection technology offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls This assay ensures efficient reproducible quantification of Eotaxin-2 a chemokine involved in immune cell chemotaxis
Catalog No. 50-000-78372 Supplier Meso Scale Discovery Supplier No. K156XQK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP Eotaxin-2 Assay is a multiplex immunoassay kit for measuring Eotaxin-2 in non-human primate samples like serum and plasma It uses MSD s electrochemiluminescent detection technology offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls This assay ensures efficient reproducible quantification of Eotaxin-2 a chemokine involved in immune cell chemotaxis