Learn More
Meso Scale Discovery UPLEX NHP Eotaxin2 ASY384well
SDP
Catalog No. 5000078459
Encompass
The Meso Scale Discovery U-PLEX NHP Eotaxin-2 384-well Assay Kit is a multiplex immunoassay that measures Eotaxin-2 levels in non-human primate serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies Read Buffer Sample Diluent and Calibrator It can test up to 384 samples in a single run and has a testing time of 3 5 hours The assay has high specificity and less than 1% cross-reactivity with other analytes
Catalog No. 50-000-78459 Supplier Meso Scale Discovery Supplier No. K256XQK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP Eotaxin-2 384-well Assay Kit is a multiplex immunoassay that measures Eotaxin-2 levels in non-human primate serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies Read Buffer Sample Diluent and Calibrator It can test up to 384 samples in a single run and has a testing time of 3 5 hours The assay has high specificity and less than 1% cross-reactivity with other analytes