Learn More
Meso Scale Discovery UPLEX NHP Eotaxin2 ASY384well
SDP
Catalog No. 5000080270
Encompass
The Meso Scale Discovery U-PLEX NHP Eotaxin-2 384-well assay kit is a multiplex immunoassay that measures Eotaxin-2 levels in non-human primate serum plasma and other biological samples This kit uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies calibrators controls 10X read buffer sample diluent and wash buffer The assay has a typical sensitivity of 0 08 pg/mL and an upper limit of quantification of 10 000 pg/mL This product enables efficient multiplexed analysis of NHP Eotaxin-2
Catalog No. 50-000-80270 Supplier Meso Scale Discovery Supplier No. K256XQK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP Eotaxin-2 384-well assay kit is a multiplex immunoassay that measures Eotaxin-2 levels in non-human primate serum plasma and other biological samples This kit uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies calibrators controls 10X read buffer sample diluent and wash buffer The assay has a typical sensitivity of 0 08 pg/mL and an upper limit of quantification of 10 000 pg/mL This product enables efficient multiplexed analysis of NHP Eotaxin-2