Learn More
Meso Scale Discovery UPLEX NHP Eotaxin3 ASY 25 PL
SDP
Catalog No. 5000079068
Encompass
The Meso Scale Discovery U-PLEX NHP Eotaxin-3 Assay Kit is a multiplex immunoassay for measuring Eotaxin-3 in non-human primate samples It uses electrochemiluminescence detection for high sensitivity and precision The kit includes a 96-well plate with capture antibodies ruthenium-labeled detection antibodies calibrators controls diluent read buffer wash buffer and seals Sensitivity is 0 08 pg/mL with a quantification limit of 2500 pg/mL enabling quantitative measurement of Eotaxin-3 a chemokine in immune cell chemotaxis
Catalog No. 50-000-79068 Supplier Meso Scale Discovery Supplier No. K156UEK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP Eotaxin-3 Assay Kit is a multiplex immunoassay for measuring Eotaxin-3 in non-human primate samples It uses electrochemiluminescence detection for high sensitivity and precision The kit includes a 96-well plate with capture antibodies ruthenium-labeled detection antibodies calibrators controls diluent read buffer wash buffer and seals Sensitivity is 0 08 pg/mL with a quantification limit of 2500 pg/mL enabling quantitative measurement of Eotaxin-3 a chemokine in immune cell chemotaxis