Learn More
Meso Scale Discovery UPLEX NHP Eotaxin3 ASY384well
SDP
Catalog No. 5000081185
Encompass
The Meso Scale Discovery U-PLEX NHP Eotaxin-3 384-well Assay Kit is a multiplex immunoassay that measures Eotaxin-3 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies Read Buffer and calibrator It requires 2 5 hours to complete and has a shelf life of 9 months when stored at 2-8 C The assay measures Eotaxin-3 levels from 0 06 pg/mL to 10 000 pg/mL
Catalog No. 50-000-81185 Supplier Meso Scale Discovery Supplier No. K256UEK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP Eotaxin-3 384-well Assay Kit is a multiplex immunoassay that measures Eotaxin-3 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies Read Buffer and calibrator It requires 2 5 hours to complete and has a shelf life of 9 months when stored at 2-8 C The assay measures Eotaxin-3 levels from 0 06 pg/mL to 10 000 pg/mL