Learn More
Meso Scale Discovery UPLEX NHP Eotaxin3 ASY384well
SDP
Supplier:  Meso Scale Discovery K256UEK2
Encompass
The Meso Scale Discovery U-PLEX NHP Eotaxin-3 384-well Assay Kit is a multiplex immunoassay that measures human Eotaxin-3 levels in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption This 384-well plate assay allows for efficient and economic profiling of large sample sets With minimal hands-on time it enables quantification of Eotaxin-3 biomarkers related to inflammation and immune response
Catalog No. 50-000-81227
Add to Cart
Request a Quote