Learn More
Meso Scale Discovery UPLEX NHP FLT3L ASY384well
SDP
Catalog No. 5000079239
Encompass
The Meso Scale Discovery U-PLEX NHP FLT3L 384-Well Assay Kit is a multiplex immunoassay that measures human FLT3 ligand in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls It allows efficient and accurate quantification of FLT3 ligand from small sample volumes The kit has high lot-to-lot consistency and is suitable for biomarker research and drug discovery applications
Catalog No. 50-000-79239 Supplier Meso Scale Discovery Supplier No. K256XFK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP FLT3L 384-Well Assay Kit is a multiplex immunoassay that measures human FLT3 ligand in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls It allows efficient and accurate quantification of FLT3 ligand from small sample volumes The kit has high lot-to-lot consistency and is suitable for biomarker research and drug discovery applications